This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other 8430430B14RIK SIRNA Assays ›
Gene Symbol:
8430430B14Rik
Gene Name:
RIKEN cDNA 8430430B14 gene
Gene Aliases:
-
Chromosome Location:
Chr.17: 83996062 - 84013419 on Build GRCm38
UniGene ID:
Mm.243918
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID n379242

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 8430430B14Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK020236.1 - 2 211

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0