This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LINC01089 SIRNA Assays ›
Gene Symbol:
LINC01089
Gene Name:
long intergenic non-protein coding RNA 1089
Gene Aliases:
-
Chromosome Location:
Chr.12: 121795267 - 121803484 on Build GRCh38
UniGene ID:
Hs.524804
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n373004

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LINC01089
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK094482.1 - 1 1053
AK095700.1 - 1 2003
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BX344455.2 - 5 837 Chr. 12 - 121795769 - 121802930 [-] on Build GRCh38
GenBankmRNA BM548548.1 - 533 Chr. - - [-] on Build
GenBankmRNA BM724477.1 - 2 541 Chr. 12 - 121795731 - 121799958 [-] on Build GRCh38
GenBankmRNA CD516035.1 - 5 825 Chr. 12 - 121795815 - 121802920 [-] on Build GRCh38
GenBankmRNA BE796247.1 - 3 605 Chr. 12 - 121795735 - 121801785 [-] on Build GRCh38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0