This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GDF7 SIRNA Assays ›
Gene Symbol:
GDF7
Gene Name:
growth differentiation factor 7
Gene Aliases:
BMP12
Chromosome Location:
Chr.2: 20666664 - 20671490 on Build GRCh38
UniGene ID:
Hs.447688
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n371265

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: GDF7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182828.3 NP_878248.2 2 9018
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA U92981.1 - 1 705 Chr. 2 - 20677809 - 20679237 [+] on Build GRCh38

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

axon guidance
positive regulation of pathway-restricted SMAD protein phosphorylation
roof plate formation
spinal cord association neuron differentiation
gland morphogenesis
BMP signaling pathway
epithelial cell differentiation
midbrain development
activin receptor signaling pathway
growth
regulation of apoptotic process
regulation of MAPK cascade
cell fate commitment
positive regulation of neuron differentiation
positive regulation of transcription, DNA-templated
cell development
reproductive structure development
branching morphogenesis of an epithelial tube
forebrain morphogenesis
pathway-restricted SMAD protein phosphorylation
SMAD protein signal transduction
morphogenesis of an epithelial fold
positive regulation of tendon cell differentiation
cytokine activity
transforming growth factor beta receptor binding
protein binding
growth factor activity
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0