This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NFIA SIRNA Assays ›
Gene Symbol:
NFIA
Gene Name:
nuclear factor I A
Gene Aliases:
CTF, NF-I/A, NF1-A, NFI-A, NFI-L
Chromosome Location:
Chr.1: 61077124 - 61462788 on Build GRCh38
UniGene ID:
Hs.740757
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n368227

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: NFIA
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011541514.2 XP_011539816.1 1 83
XM_017001362.1 XP_016856851.1 11
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AK092769.1 - 1 88 Chr. 1 - 61077119 - 61079248 [+] on Build GRCh38

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
DNA replication
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
viral genome replication
positive regulation of transcription from RNA polymerase II promoter
synapse maturation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription factor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0