siRNA Details
Gene Symbol: FP2234 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF447871.1 | - | 1 | 1760 | ||
AK091154.1 | - | 1 | 3760 |

Gene Symbol: FP2234 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF447871.1 | - | 1 | 1760 | ||
AK091154.1 | - | 1 | 3760 |