This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HMGCS1 SIRNA Assays ›
Gene Symbol:
HMGCS1
Gene Name:
3-hydroxy-3-methylglutaryl-CoA synthase 1
Gene Aliases:
HMGCS
Chromosome Location:
Chr.5: 43287470 - 43313512 on Build GRCh38
UniGene ID:
Hs.397729
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n360705

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: HMGCS1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098272.2 NP_001091742.1 11 5093
NM_001324219.1 NP_001311148.1 10 5080
NM_001324220.1 NP_001311149.1 11 5113
NM_001324222.1 NP_001311151.1 10 4967
NM_001324223.1 NP_001311152.1 9 4908
NM_001324224.1 NP_001311153.1 10 4987
NM_002130.7 NP_002121.4 10 5034
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BQ189002.1 - 1 406 Chr. 5 - 43287601 - 43288232 [-] on Build GRCh38
GenBankmRNA CR605684.1 - 1 1528 Chr. 5 - 43287699 - 43289354 [-] on Build GRCh38

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

liver development
lipid metabolic process
cholesterol biosynthetic process
brain development
isoprenoid biosynthetic process
male gonad development
response to low light intensity stimulus
response to purine-containing compound
response to vitamin E
response to drug
response to tellurium ion
response to lipoprotein particle
cellular response to follicle-stimulating hormone stimulus
cellular response to cholesterol
hydroxymethylglutaryl-CoA synthase activity
drug binding
isomerase activity
protein homodimerization activity
organic acid binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0