siRNA Details
Gene Symbol: LOC100506274 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038432.1 | 6 | 657 | |||
GenBank mRNA | AK093290.1 | - | 8 | 1956 | ||
BC043509.1 | - | 6 | 647 |

Gene Symbol: LOC100506274 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038432.1 | 6 | 657 | |||
GenBank mRNA | AK093290.1 | - | 8 | 1956 | ||
BC043509.1 | - | 6 | 647 |