This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LOC100506274 SIRNA Assays ›
Gene Symbol:
LOC100506274
Gene Name:
uncharacterized LOC100506274
Gene Aliases:
-
Chromosome Location:
Chr.2: 7421261 - 7450254 on Build GRCh38
UniGene ID:
Hs.177930
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n350222

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC100506274
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_038432.1 1 35
GenBank mRNA AK093290.1 - 1 32
BC043509.1 - 1 25
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BI520461.1 - 1 25 Chr. 2 - 7421271 - 7450059 [+] on Build GRCh38
GenBankmRNA DB457547.1 - 1 35 Chr. 2 - 7421261 - 7436097 [+] on Build GRCh38
GenBankmRNA DB029885.1 - 1 32 Chr. 2 - 7421265 - 7428777 [+] on Build GRCh38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0