This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PITPNC1 SIRNA Assays ›
Gene Symbol:
PITPNC1
Gene Name:
phosphatidylinositol transfer protein, cytoplasmic 1
Gene Aliases:
M-RDGB-beta, MRDGBbeta, RDGB-BETA, RDGBB, RDGBB1
Chromosome Location:
Chr.17: 67377281 - 67697263 on Build GRCh38
UniGene ID:
Hs.591185
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n341085

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PITPNC1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012417.3 NP_036549.2 9 4112
NM_181671.2 NP_858057.1 10 4231
XM_005257216.2 XP_005257273.1 9 3612
XM_017024442.1 XP_016879931.1 9 3223
XM_017024443.1 XP_016879932.1 10 3342
XM_017024444.1 XP_016879933.1 10 3733
XM_017024445.1 XP_016879934.1 10 3355
GenBank mRNA AK094724.1 - 1 66

Back To Top

More Information


Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
phospholipid transport
protein binding
lipid binding
phosphatidylinositol transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0