This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TRMT2B SIRNA Assays ›
Gene Symbol:
TRMT2B
Gene Name:
tRNA methyltransferase 2 homolog B
Gene Aliases:
CXorf34, dJ341D10.3
Chromosome Location:
Chr.X: 100973358 - 101052118 on Build GRCh38
UniGene ID:
Hs.496501
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n339464

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TRMT2B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005262195.2 XP_005262252.1 15 5344
XM_005262196.2 XP_005262253.1 13 5139
XM_005262201.2 XP_005262258.1 11 4719
XM_006724705.2 XP_006724768.1 14 5283
XM_011531042.2 XP_011529344.1 11 4837
XM_011531044.2 XP_011529346.1 13 4898
XM_017029858.1 XP_016885347.1 15 5729
XM_017029859.1 XP_016885348.1 14 5044
XM_017029860.1 XP_016885349.1 15 5343
XM_017029861.1 XP_016885350.1 14 5253
XM_017029862.1 XP_016885351.1 13 5481
XM_017029863.1 XP_016885352.1 15 5153
XM_017029864.1 XP_016885353.1 13 5345
XM_017029865.1 XP_016885354.1 13 4973
XM_017029866.1 XP_016885355.1 14 5036
XM_017029867.1 XP_016885356.1 14 5073
XM_017029868.1 XP_016885357.1 13 4946
XM_017029869.1 XP_016885358.1 12 5290
XM_017029870.1 XP_016885359.1 12 4880
XM_017029871.1 XP_016885360.1 12 4862
XM_017029872.1 XP_016885361.1 14 5086
XM_017029873.1 XP_016885362.1 14 5061
XM_017029874.1 XP_016885363.1 13 5403
XM_017029875.1 XP_016885364.1 12 4950
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AK091722.1 - 1 693 Chr. X - 101003832 - 101007973 [-] on Build GRCh38

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

RNA methylation
tRNA processing
protein binding
S-adenosylmethionine-dependent tRNA (m5U54) methyltransferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0