This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ABCB9 SIRNA Assays ›
Gene Symbol:
ABCB9
Gene Name:
ATP binding cassette subfamily B member 9
Gene Aliases:
EST122234, TAPL
Chromosome Location:
Chr.12: 122918708 - 122975160 on Build GRCh38
UniGene ID:
Hs.511951
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n338123

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ABCB9
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011538098.2 XP_011536400.1 1 1666
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AF461897.1 - 1 713 Chr. 12 - 122951241 - 122952311 [-] on Build GRCh38

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

antigen processing and presentation of peptide antigen via MHC class I
antigen processing and presentation of exogenous protein antigen via MHC class Ib, TAP-dependent
antigen processing and presentation of endogenous peptide antigen via MHC class I via ER pathway, TAP-dependent
antigen processing and presentation of endogenous peptide antigen via MHC class Ib via ER pathway, TAP-dependent
positive regulation of antigen processing and presentation of peptide antigen via MHC class I
protein transport
peptide transport
oligopeptide transmembrane transport
transmembrane transport
protein binding
ATP binding
oligopeptide-transporting ATPase activity
peptide-transporting ATPase activity
substrate-specific transmembrane transporter activity
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0