This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other C12ORF40 SIRNA Assays ›
Gene Symbol:
C12orf40
Gene Name:
chromosome 12 open reading frame 40
Gene Aliases:
HEL-206, HEL-S-94
Chromosome Location:
Chr.12: 39626167 - 39908300 on Build GRCh38
UniGene ID:
Hs.648205
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n336922

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C12orf40
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_135051.1 19 3063
GenBank mRNA AK097445.1 - 19 3010
EU668334.1 - 19 3010
GQ472216.1 - 19 3010

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0