siRNA Details
Gene Symbol: LOC101929124 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110427.1 | 5 | 3306 | |||
GenBank mRNA | AK126236.1 | - | 5 | 3308 |

Gene Symbol: LOC101929124 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110427.1 | 5 | 3306 | |||
GenBank mRNA | AK126236.1 | - | 5 | 3308 |