This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LOC100128401 SIRNA Assays ›
Gene Symbol:
LOC100128401 HPS4
Gene Name:
uncharacterized LOC100128401
HPS4, biogenesis of lysosomal organelles complex 3 subunit 2
Gene Aliases:
-
BLOC3S2, LE
Chromosome Location:
-
Chromosome Location:
Chr.22: 26443613 - 26483863 on Build GRCh38
UniGene ID:
Hs.567547
Hs.474436
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID n330343

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC100128401
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK124497.1 - 1 2273
Gene Symbol: HPS4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011530492.1 XP_011528794.1 16 2856
XM_017029045.1 XP_016884534.1 15 2802
XM_017029048.1 XP_016884537.1 14 2724
XM_017029049.1 XP_016884538.1 14 2629

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein targeting
lysosome organization
blood coagulation
hemostasis
melanocyte differentiation
positive regulation of eye pigmentation
protein stabilization
positive regulation of protein targeting to mitochondrion
protein binding
protein homodimerization activity
protein dimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0