This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other STMN3 SIRNA Assays ›
Gene Symbol:
STMN3
Gene Name:
stathmin 3
Gene Aliases:
SCLIP
Chromosome Location:
Chr.20: 63639705 - 63653610 on Build GRCh38
UniGene ID:
Hs.639609
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n328596

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: STMN3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK131326.1 - 1 154
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CR613563.1 - 1 178 Chr. 20 - 63639845 - 63641845 [-] on Build GRCh38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0