siRNA Details
Gene Symbol: LOC101928730 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120422.1 | 1 | 717 | |||
GenBank mRNA | BC034786.1 | - | 1 | 717 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI828502.1 | - | 1 | 717 | Chr. 13 - 112271488 - 112273000 [-] on Build GRCh38 |
