This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF691 SIRNA Assays ›
Gene Symbol:
ZNF691
Gene Name:
zinc finger protein 691
Gene Aliases:
Zfp691
Chromosome Location:
Chr.1: 42846573 - 42852477 on Build GRCh38
UniGene ID:
Hs.20879
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n312525

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF691
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017001401.1 XP_016856890.1 1 1744
XM_017001403.1 XP_016856892.1 1 1743
GenBank mRNA AK095184.1 - 1 1802
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CB105122.1 - 1 103 Chr. 1 - 42848316 - 42848812 [+] on Build GRCh38

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
RNA polymerase II regulatory region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header