This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NCBP3 SIRNA Assays ›
Gene Symbol:
NCBP3
Gene Name:
nuclear cap binding subunit 3
Gene Aliases:
C17orf85, ELG, HSA277841
Chromosome Location:
Chr.17: 3806748 - 3846251 on Build GRCh38
UniGene ID:
Hs.120963
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n308783

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: NCBP3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114118.2 NP_001107590.1 13 7898
XM_005256717.2 XP_005256774.1 14 7436
GenBank mRNA AK056724.1 - 1 1818

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

7-methylguanosine mRNA capping
mRNA transport
defense response to virus
nucleotide binding
RNA 7-methylguanosine cap binding
mRNA binding
protein binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0