siRNA Details
Gene Symbol: LOC645967 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134652.1 | 1 | 991 | |||
GenBank mRNA | AB593152.1 | - | 1 | 991 | ||
CA395838.1 | - | 1 | 247 |

Gene Symbol: LOC645967 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134652.1 | 1 | 991 | |||
GenBank mRNA | AB593152.1 | - | 1 | 991 | ||
CA395838.1 | - | 1 | 247 |