This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PKD1L1 SIRNA Assays ›
Gene Symbol:
PKD1L1
Gene Name:
polycystin 1 like 1, transient receptor potential channel interacting
Gene Aliases:
PRO19563
Chromosome Location:
Chr.7: 47774603 - 47958622 on Build GRCh38
UniGene ID:
Hs.195979
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n297731

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PKD1L1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017011798.1 XP_016867287.1 1 2423
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BE888478.1 - 1 553 Chr. 7 - 47956038 - 47956751 [-] on Build GRCh38
GenBankmRNA CF145720.1 - 1 526 Chr. 7 - 47955973 - 47956718 [-] on Build GRCh38
GenBankmRNA BQ548643.1 - 1 324 Chr. 7 - 47956000 - 47956523 [-] on Build GRCh38
GenBankmRNA DB484785.1 - 1 359 Chr. 7 - 47956122 - 47956556 [-] on Build GRCh38
GenBankmRNA AK001279.1 - 1 1114 Chr. 7 - 47955352 - 47957314 [-] on Build GRCh38
GenBankmRNA DB504382.1 - 1 357 Chr. 7 - 47956092 - 47956556 [-] on Build GRCh38
GenBankmRNA CB963771.1 - 1 463 Chr. 7 - 47955970 - 47956660 [-] on Build GRCh38
GenBankmRNA CF145705.1 - 1 526 Chr. 7 - 47956042 - 47956718 [-] on Build GRCh38
GenBankmRNA DB488127.1 - 1 357 Chr. 7 - 47956111 - 47956556 [-] on Build GRCh38
GenBankmRNA CN302055.1 - 1 381 Chr. 7 - 47955831 - 47956580 [-] on Build GRCh38

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

detection of nodal flow
single organismal cell-cell adhesion
detection of mechanical stimulus
calcium ion transmembrane transport
left/right axis specification
calcium channel activity
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header