This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZFP69B SIRNA Assays ›
Gene Symbol:
ZFP69B
Gene Name:
ZFP69 zinc finger protein B
Gene Aliases:
ZKSCAN23B, ZNF643, ZSCAN54B
Chromosome Location:
Chr.1: 40450068 - 40466765 on Build GRCh38
UniGene ID:
Hs.133034
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n295988

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZFP69B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005271137.2 XP_005271194.1 6 4273
XM_005271139.3 XP_005271196.1 5 4221
GenBank mRNA AK094134.1 - 1 1559
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CB114546.1 - 1 357 Chr. 1 - 40465521 - 40466152 [+] on Build GRCh38
GenBankmRNA DA517331.1 - 1 195 Chr. 1 - 40465684 - 40466243 [+] on Build GRCh38
GenBankmRNA DA801448.1 - 1 503 Chr. 1 - 40465375 - 40465946 [+] on Build GRCh38

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0