siRNA Details
Gene Symbol: LOC101928988 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120344.1 | 4 | 785 | |||
GenBank mRNA | BC039388.1 | - | 4 | 785 |

Gene Symbol: LOC101928988 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120344.1 | 4 | 785 | |||
GenBank mRNA | BC039388.1 | - | 4 | 785 |