This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FBXO31 SIRNA Assays ›
Gene Symbol:
FBXO31
Gene Name:
F-box protein 31
Gene Aliases:
FBX14, FBXO14, Fbx31, MRT45, pp2386
Chromosome Location:
Chr.16: 87326987 - 87392107 on Build GRCh38
UniGene ID:
Hs.567582
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n295385

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FBXO31
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282683.1 NP_001269612.1 10 5344
NM_024735.4 NP_079011.3 9 5596
GenBank mRNA BX106211.1 - 1 489
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CD251462.1 - 378 Chr. - - [-] on Build
GenBankmRNA BM694819.1 - 1 26 Chr. 16 - 87326988 - 87327386 [-] on Build GRCh38
GenBankmRNA CR621785.1 - 1 2028 Chr. 16 - 87327016 - 87329388 [-] on Build GRCh38

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
cellular response to DNA damage stimulus
negative regulation of cyclin-dependent protein serine/threonine kinase by cyclin degradation
SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
mitotic G1 DNA damage checkpoint
positive regulation of dendrite morphogenesis
positive regulation of neuron migration
ubiquitin-protein transferase activity
cyclin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0