This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PLXDC2 SIRNA Assays ›
Gene Symbol:
PLXDC2
Gene Name:
plexin domain containing 2
Gene Aliases:
TEM7R
Chromosome Location:
Chr.10: 19816215 - 20289856 on Build GRCh38
UniGene ID:
Hs.658134
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n295212

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PLXDC2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282736.1 NP_001269665.1 13 4642
NM_032812.8 NP_116201.7 14 4789
GenBank mRNA AK127644.1 - 1 1367

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header