This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC9A3 SIRNA Assays ›
Gene Symbol:
SLC9A3
Gene Name:
solute carrier family 9 member A3
Gene Aliases:
DIAR8, NHE-3, NHE3
Chromosome Location:
Chr.5: 471986 - 524434 on Build GRCh38
UniGene ID:
Hs.658120
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n292742

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SLC9A3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001284351.2 NP_001271280.1 17 3540
NM_004174.3 NP_004165.2 17 3567
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CN360410.1 - 1 465 Chr. 5 - 472233 - 472884 [-] on Build GRCh38
GenBankmRNA DA525226.1 - 1 549 Chr. 5 - 472418 - 472977 [-] on Build GRCh38
GenBankmRNA BM450275.1 - 1 503 Chr. 5 - 471984 - 472931 [-] on Build GRCh38
GenBankmRNA BI917207.1 - 1 478 Chr. 5 - 472340 - 472901 [-] on Build GRCh38
GenBankmRNA BM807725.1 - 608 Chr. - - [-] on Build
GenBankmRNA BI913040.1 - 1 565 Chr. 5 - 472272 - 472994 [-] on Build GRCh38
GenBankmRNA HY118290.1 - 1 540 Chr. 5 - 472416 - 472967 [-] on Build GRCh38
GenBankmRNA DA770151.1 - 1 537 Chr. 5 - 472394 - 472965 [-] on Build GRCh38
GenBankmRNA DA605548.1 - 1 579 Chr. 5 - 472382 - 473007 [-] on Build GRCh38
GenBankmRNA DN999780.1 - 1 632 Chr. 5 - 472414 - 473052 [-] on Build GRCh38
GenBankmRNA DA579043.1 - 1 539 Chr. 5 - 472408 - 472967 [-] on Build GRCh38
GenBankmRNA BU159298.1 - 547 Chr. - - [-] on Build
GenBankmRNA AK023178.1 - 1 635 Chr. 5 - 470511 - 473063 [-] on Build GRCh38
GenBankmRNA DA297367.1 - 1 559 Chr. 5 - 472412 - 472986 [-] on Build GRCh38
GenBankmRNA AL556583.3 - 1 620 Chr. 5 - 472180 - 473049 [-] on Build GRCh38
GenBankmRNA BI546950.1 - 1 533 Chr. 5 - 472230 - 472956 [-] on Build GRCh38
GenBankmRNA BI917212.1 - 1 524 Chr. 5 - 472354 - 472952 [-] on Build GRCh38

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of sodium ion transport
ion transport
receptor-mediated endocytosis
circadian rhythm
response to glucocorticoid
regulation of intracellular pH
potassium ion transmembrane transport
sodium ion import across plasma membrane
hydrogen ion transmembrane transport
protein binding
sodium:proton antiporter activity
potassium:proton antiporter activity
PDZ domain binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0