This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZSCAN31 SIRNA Assays ›
Gene Symbol:
ZSCAN31
Gene Name:
zinc finger and SCAN domain containing 31
Gene Aliases:
ZNF20-Lp, ZNF310P, ZNF323
Chromosome Location:
Chr.6: 28324737 - 28358202 on Build GRCh38
UniGene ID:
Hs.656413
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n291218

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZSCAN31
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005249296.4 XP_005249353.1 1 3008
XM_011514807.2 XP_011513109.1 1 3008
XM_011514808.2 XP_011513110.1 1 3008
XM_017011196.1 XP_016866685.1 1 3008
GenBank mRNA AK123280.1 - 1 1077

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0