This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NKTR SIRNA Assays ›
Gene Symbol:
NKTR
Gene Name:
natural killer cell triggering receptor
Gene Aliases:
p104
Chromosome Location:
Chr.3: 42600655 - 42648741 on Build GRCh38
UniGene ID:
Hs.529509
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n284356

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: NKTR
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011533747.2 XP_011532049.1 8 540
XM_017006476.1 XP_016861965.1 2 1734
GenBank mRNA AB209888.1 - 6 2459
BX538094.1 - 2507
BX641127.1 - 1 50
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DB109227.1 - 1 312 Chr. 3 - 42626924 - 42627498 [+] on Build GRCh38
GenBankmRNA AK093752.1 - 1 312 Chr. 3 - 42626924 - 42629010 [+] on Build GRCh38
GenBankmRNA DA817093.1 - 1 420 Chr. 3 - 42626817 - 42627345 [+] on Build GRCh38
GenBankmRNA DA802958.1 - 1 276 Chr. 3 - 42626960 - 42627481 [+] on Build GRCh38

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: AK163916, AK052484.

Gene Ontology Categories:

Function(s) Process(es)

protein peptidyl-prolyl isomerization
protein folding
peptidyl-prolyl cis-trans isomerase activity
cyclosporin A binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header