This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GTF3C4 SIRNA Assays ›
Gene Symbol:
GTF3C4
Gene Name:
general transcription factor IIIC subunit 4
Gene Aliases:
KAT12, TF3C-delta, TFIII90, TFIIIC290, TFIIIC90, TFIIICDELTA
Chromosome Location:
Chr.9: 132670035 - 132694955 on Build GRCh38
UniGene ID:
Hs.22302
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n284119

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: GTF3C4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012204.3 NP_036336.2 5 4805
NR_133925.1 6 4843
XM_017015318.1 XP_016870807.1 5 2588
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BF980709.1 - 194 Chr. - - [-] on Build
GenBankmRNA AK126436.1 - 1 2240 Chr. 9 - 132688475 - 132690801 [+] on Build GRCh38
GenBankmRNA DB281620.1 - 1 430 Chr. 9 - 132690285 - 132690845 [+] on Build GRCh38
GenBankmRNA BI060228.1 - 1 201 Chr. 9 - 132690537 - 132690787 [+] on Build GRCh38
GenBankmRNA CN430666.1 - 1 186 Chr. 9 - 132690532 - 132690983 [+] on Build GRCh38
GenBankmRNA DA758514.1 - 2 436 Chr. 9 - 132670497 - 132690810 [+] on Build GRCh38
GenBankmRNA CN430665.1 - 1 500 Chr. 9 - 132690218 - 132690924 [+] on Build GRCh38
GenBankmRNA BG112922.1 - 1 455 Chr. 9 - 132690263 - 132690812 [+] on Build GRCh38
GenBankmRNA DA567668.1 - 1 204 Chr. 9 - 132690514 - 132691067 [+] on Build GRCh38

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
transcription from RNA polymerase III promoter
transcription initiation from RNA polymerase III promoter
histone acetylation
5S class rRNA transcription from RNA polymerase III type 1 promoter
tRNA transcription from RNA polymerase III promoter
positive regulation of catalytic activity
DNA binding
histone acetyltransferase activity
protein binding
enzyme activator activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0