This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF75D SIRNA Assays ›
Gene Symbol:
ZNF75D
Gene Name:
zinc finger protein 75D
Gene Aliases:
D8C6, ZKSCAN24, ZNF75, ZNF82, ZSCAN28
Chromosome Location:
Chr.X: 135248589 - 135344109 on Build GRCh38
UniGene ID:
Hs.371717
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n281897

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF75D
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_110381.1 3 1179
GenBank mRNA AY730281.1 - 2 372
BC029787.1 - 3 1179
BG186944.1 - 2 198

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
transcription factor activity, sequence-specific DNA binding
zinc ion binding
sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0