siRNA Details
Gene Symbol: LOC101928372 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110695.1 | 1 | 103 | |||
GenBank mRNA | BC035240.1 | - | 1 | 103 |

Gene Symbol: LOC101928372 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110695.1 | 1 | 103 | |||
GenBank mRNA | BC035240.1 | - | 1 | 103 |