This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LOC100128727 SIRNA Assays ›
Gene Symbol:
LOC100128727 DGKI
Gene Name:
uncharacterized LOC100128727
diacylglycerol kinase iota
Gene Aliases:
-
DGK-IOTA
Chromosome Location:
-
Chromosome Location:
Chr.7: 137384283 - 137846864 on Build GRCh38
UniGene ID:
Hs.636145
Hs.242947
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID n279549

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC100128727
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK124397.1 - 1 3369
Gene Symbol: DGKI
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011516684.2 XP_011514986.2 27 17183
XM_017012780.1 XP_016868269.1 34 10000
XM_017012781.1 XP_016868270.1 34 9984
XM_017012782.1 XP_016868271.1 33 9885
XM_017012783.1 XP_016868272.1 33 9876
XM_017012784.1 XP_016868273.1 32 9810
XM_017012785.1 XP_016868274.1 32 10187
XM_017012786.1 XP_016868275.1 34 9687
XM_017012787.1 XP_016868276.1 33 9552
XM_017012788.1 XP_016868277.1 34 9670
XM_017012789.1 XP_016868278.1 20 8396

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein kinase C-activating G-protein coupled receptor signaling pathway
neurotransmitter secretion
phosphorylation
platelet activation
negative regulation of GTPase activity
intracellular signal transduction
positive regulation of Ras protein signal transduction
habituation
regulation of synaptic transmission, glutamatergic
excitatory postsynaptic potential
regulation of long term synaptic depression
diacylglycerol kinase activity
GTPase inhibitor activity
protein binding
ATP binding
Ras GTPase binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header