This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CNOT7 SIRNA Assays ›
Gene Symbol:
CNOT7
Gene Name:
CCR4-NOT transcription complex subunit 7
Gene Aliases:
CAF-1, CAF1, Caf1a, hCAF-1
Chromosome Location:
Chr.8: 17226030 - 17246909 on Build GRCh38
UniGene ID:
Hs.587465
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n276398

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CNOT7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005273481.2 XP_005273538.1 8 5167
GenBank mRNA AK090522.1 - 1 1255
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AK091357.1 - 1 431 Chr. 8 - 17224967 - 17227277 [-] on Build GRCh38

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

nuclear-transcribed mRNA poly(A) tail shortening
deadenylation-dependent decapping of nuclear-transcribed mRNA
carbohydrate metabolic process
transcription, DNA-templated
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
signal transduction
positive regulation of cell proliferation
negative regulation of cell proliferation
negative regulation of gene expression
gene silencing by RNA
cytoplasmic mRNA processing body assembly
gene silencing by miRNA
exonucleolytic nuclear-transcribed mRNA catabolic process involved in deadenylation-dependent decay
positive regulation of transcription from RNA polymerase II promoter
positive regulation of nuclear-transcribed mRNA poly(A) tail shortening
positive regulation of mRNA catabolic process
RNA phosphodiester bond hydrolysis, exonucleolytic
positive regulation of nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
3'-5'-exoribonuclease activity
transcription factor activity, sequence-specific DNA binding
RNA binding
exoribonuclease activity
poly(A)-specific ribonuclease activity
signal transducer activity
protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0