siRNA Details
Gene Symbol: LINC00327 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AY927579.1 | - | LIT3578 AY927579 |
1 | 1061 |

Gene Symbol: LINC00327 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AY927579.1 | - | LIT3578 AY927579 |
1 | 1061 |