This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SNHG5 SIRNA Assays ›
Gene Symbol:
SNHG5
Gene Name:
small nucleolar RNA host gene 5
Gene Aliases:
C6orf160, LINC00044, NCRNA00044, U50HG, bA33E24.2
Chromosome Location:
Chr.6: 85677007 - 85678733 on Build GRCh38
UniGene ID:
Hs.292457
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n272219

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SNHG5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003038.2 1, 2 37
GenBank mRNA CD384894.1 - 1, 2 54
CD388674.1 - 1, 2 54
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CD389879.1 - 1, 2 60 Chr. 6 - 85677079 - 85678733 [-] on Build GRCh38
GenBankmRNA HY031639.1 - 1, 2 36 Chr. 6 - 85677084 - 85678732 [-] on Build GRCh38
GenBankmRNA CD249518.1 - 1, 2 63 Chr. 6 - 85677079 - 85678733 [-] on Build GRCh38
GenBankmRNA BM920267.1 - 20 Chr. - - [-] on Build
GenBankmRNA F19304.2 - 21 Chr. 11 - 8244443 - 8244457 [+] on Build GRCh38
GenBankmRNA CD177169.1 - 1, 2 59 Chr. 6 - 85677082 - 85678733 [-] on Build GRCh38
GenBankmRNA CN368117.1 - 1, 2 29 Chr. 6 - 85677007 - 85678717 [-] on Build GRCh38
GenBankmRNA EB388820.1 - 1, 2 26 Chr. 6 - 85677082 - 85678722 [-] on Build GRCh38
GenBankmRNA BG333157.1 - 1, 2 28 Chr. 6 - 85677084 - 85678723 [-] on Build GRCh38
GenBankmRNA BQ432969.1 - 1, 2 46 Chr. 6 - 85677082 - 85678719 [-] on Build GRCh38
GenBankmRNA CD385963.1 - 1, 2 59 Chr. 6 - 85677082 - 85678731 [-] on Build GRCh38
GenBankmRNA CD174499.1 - 1, 2 59 Chr. 6 - 85677082 - 85678733 [-] on Build GRCh38
GenBankmRNA BF209379.1 - 1, 2 38 Chr. 6 - 85677078 - 85678733 [-] on Build GRCh38
GenBankmRNA DA482879.1 - 1, 2 37 Chr. 6 - 85677079 - 85678733 [-] on Build GRCh38
GenBankmRNA CD175099.1 - 1, 2 59 Chr. 6 - 85677082 - 85678733 [-] on Build GRCh38
GenBankmRNA CD520230.1 - 1, 2 40 Chr. 6 - 85677083 - 85678736 [-] on Build GRCh38
GenBankmRNA BU532076.1 - 40 Chr. - - [-] on Build
GenBankmRNA BU182289.1 - 1, 2 23 Chr. 6 - 85677082 - 85678719 [-] on Build GRCh38
GenBankmRNA BM695213.1 - 1, 2 19 Chr. 6 - 85677082 - 85678715 [-] on Build GRCh38
GenBankmRNA BU531826.1 - 1, 2 40 Chr. 6 - 85677082 - 85678736 [-] on Build GRCh38
GenBankmRNA BQ230279.1 - 18 Chr. - - [-] on Build
GenBankmRNA AW972834.1 - 131 Chr. - - [-] on Build
GenBankmRNA DB449890.1 - 1, 2 40 Chr. 6 - 85677082 - 85678736 [-] on Build GRCh38
GenBankmRNA CD249906.1 - 1, 2 43 Chr. 6 - 85677085 - 85678731 [-] on Build GRCh38
GenBankmRNA BU954986.1 - 40 Chr. - - [-] on Build
GenBankmRNA BF971639.1 - 21 Chr. - - [-] on Build
GenBankmRNA BF678689.1 - 38 Chr. - - [-] on Build
GenBankmRNA BQ876776.1 - 17 Chr. - - [-] on Build
GenBankmRNA CD175168.1 - 1, 2 59 Chr. 6 - 85677082 - 85678733 [-] on Build GRCh38
GenBankmRNA CD389769.1 - 1, 2 56 Chr. 6 - 85677082 - 85678733 [-] on Build GRCh38
GenBankmRNA CD175213.1 - 1, 2 53 Chr. 6 - 85677085 - 85678733 [-] on Build GRCh38
GenBankmRNA F30675.1 - 1, 2 19 Chr. 6 - 85677439 - 85678715 [-] on Build GRCh38
GenBankmRNA CD048591.1 - 59 Chr. - - [-] on Build
GenBankmRNA CD048947.1 - 60 Chr. - - [-] on Build
GenBankmRNA CD388186.1 - 1, 2 57 Chr. 6 - 85677079 - 85678733 [-] on Build GRCh38
GenBankmRNA CD177114.1 - 1, 2 59 Chr. 6 - 85677079 - 85678733 [-] on Build GRCh38
GenBankmRNA CD247795.1 - 1, 2 56 Chr. 6 - 85677085 - 85678733 [-] on Build GRCh38
GenBankmRNA BU532664.1 - 1, 2 40 Chr. 6 - 85677085 - 85678731 [-] on Build GRCh38
GenBankmRNA CD177483.1 - 1, 2 53 Chr. 6 - 85677085 - 85678733 [-] on Build GRCh38
GenBankmRNA CN368120.1 - 1, 2 29 Chr. 6 - 85677082 - 85678717 [-] on Build GRCh38
GenBankmRNA CD176393.1 - 1, 2 63 Chr. 6 - 85677079 - 85678733 [-] on Build GRCh38
GenBankmRNA BU536013.1 - 1, 2 40 Chr. 6 - 85677082 - 85678736 [-] on Build GRCh38
GenBankmRNA BU588624.1 - 1, 2 40 Chr. 6 - 85677082 - 85678736 [-] on Build GRCh38
GenBankmRNA CD249877.1 - 1, 2 60 Chr. 6 - 85677079 - 85678725 [-] on Build GRCh38
GenBankmRNA CD385054.1 - 63 Chr. - - [-] on Build
GenBankmRNA CD388557.1 - 1, 2 56 Chr. 6 - 85677079 - 85678733 [-] on Build GRCh38
GenBankmRNA CD250044.1 - 1, 2 65 Chr. 6 - 85677082 - 85678734 [-] on Build GRCh38
GenBankmRNA CD049899.1 - 55 Chr. - - [-] on Build
GenBankmRNA BF307294.1 - 15 Chr. - - [-] on Build
GenBankmRNA CD175102.1 - 1, 2 58 Chr. 6 - 85677079 - 85678733 [-] on Build GRCh38
GenBankmRNA CD390262.1 - 1, 2 53 Chr. 6 - 85677085 - 85678731 [-] on Build GRCh38
GenBankmRNA CD248963.1 - 60 Chr. - - [-] on Build
GenBankmRNA CB962559.1 - 1, 2 40 Chr. 6 - 85677079 - 85678736 [-] on Build GRCh38
GenBankmRNA EL947347.1 - 1, 2 25 Chr. 6 - 85677077 - 85678719 [-] on Build GRCh38
GenBankmRNA CB128964.1 - 1, 2 37 Chr. 6 - 85677082 - 85678733 [-] on Build GRCh38
GenBankmRNA BG759735.1 - 1, 2 20 Chr. 6 - 85677082 - 85678716 [-] on Build GRCh38
GenBankmRNA HY168432.1 - 1, 2 36 Chr. 6 - 85677083 - 85678732 [-] on Build GRCh38
GenBankmRNA CD389793.1 - 58 Chr. - - [-] on Build
GenBankmRNA BF966370.2 - 1, 2 43 Chr. 6 - 85677082 - 85678737 [-] on Build GRCh38
GenBankmRNA BM840935.1 - 1, 2 59 Chr. 6 - 85677082 - 85678733 [-] on Build GRCh38
GenBankmRNA CD173592.1 - 1, 2 63 Chr. 6 - 85677082 - 85678733 [-] on Build GRCh38
GenBankmRNA BG330935.1 - 1, 2 28 Chr. 6 - 85677079 - 85678723 [-] on Build GRCh38
GenBankmRNA CD387585.1 - 52 Chr. - - [-] on Build
GenBankmRNA BU588037.1 - 40 Chr. - - [-] on Build
GenBankmRNA W04663.1 - 19 Chr. - - [-] on Build
GenBankmRNA BF980513.1 - 14 Chr. - - [-] on Build
GenBankmRNA BF218089.1 - 1, 2 41 Chr. 6 - 85677079 - 85678736 [-] on Build GRCh38
GenBankmRNA CD173436.1 - 1, 2 59 Chr. 6 - 85677082 - 85678733 [-] on Build GRCh38
GenBankmRNA CB117414.1 - 1, 2 16 Chr. 6 - 85677082 - 85678712 [-] on Build GRCh38
GenBankmRNA CD175427.1 - 1, 2 59 Chr. 6 - 85677082 - 85678733 [-] on Build GRCh38
GenBankmRNA AA385186.1 - 1, 2 15 Chr. 6 - 85677169 - 85678711 [-] on Build GRCh38
GenBankmRNA CD177627.1 - 1, 2 64 Chr. 6 - 85677082 - 85678734 [-] on Build GRCh38
GenBankmRNA BG332327.1 - 21 Chr. - - [-] on Build
GenBankmRNA BG705462.1 - 1, 2 43 Chr. 6 - 85677082 - 85678737 [-] on Build GRCh38
GenBankmRNA BI090148.1 - 21 Chr. - - [-] on Build
GenBankmRNA AA308290.1 - 1, 2 13 Chr. 6 - 85677083 - 85678709 [-] on Build GRCh38
GenBankmRNA AJ714332.1 - 1, 2 30 Chr. 6 - 85677100 - 85678715 [-] on Build GRCh38
GenBankmRNA CD175562.1 - 1, 2 59 Chr. 6 - 85677082 - 85678733 [-] on Build GRCh38
GenBankmRNA BU602968.1 - 40 Chr. - - [-] on Build
GenBankmRNA CD172948.1 - 1, 2 56 Chr. 6 - 85677079 - 85678733 [-] on Build GRCh38
GenBankmRNA CD176074.1 - 1, 2 62 Chr. 6 - 85677085 - 85678734 [-] on Build GRCh38
GenBankmRNA CD175083.1 - 1, 2 55 Chr. 6 - 85677079 - 85678733 [-] on Build GRCh38
GenBankmRNA HY027691.1 - 1, 2 36 Chr. 6 - 85677083 - 85678732 [-] on Build GRCh38
GenBankmRNA HY101068.1 - 1, 2 36 Chr. 6 - 85677079 - 85678732 [-] on Build GRCh38
GenBankmRNA CD048516.1 - 94 Chr. - - [-] on Build
GenBankmRNA CD175206.1 - 1, 2 65 Chr. 6 - 85677077 - 85678733 [-] on Build GRCh38
GenBankmRNA BG754036.1 - 1, 2 20 Chr. 6 - 85677082 - 85678716 [-] on Build GRCh38
GenBankmRNA CD174861.1 - 1, 2 60 Chr. 6 - 85677082 - 85678733 [-] on Build GRCh38
GenBankmRNA CD177343.1 - 1, 2 60 Chr. 6 - 85677085 - 85678732 [-] on Build GRCh38
GenBankmRNA BM763888.1 - 1, 2 37 Chr. 6 - 85677082 - 85678733 [-] on Build GRCh38
GenBankmRNA CD675327.1 - 1, 2 33 Chr. 6 - 85677082 - 85678720 [-] on Build GRCh38
GenBankmRNA BU941995.1 - 40 Chr. - - [-] on Build
GenBankmRNA CD173371.1 - 1, 2 63 Chr. 6 - 85677082 - 85678733 [-] on Build GRCh38
GenBankmRNA BG758157.1 - 1, 2 22 Chr. 6 - 85677082 - 85678717 [-] on Build GRCh38
GenBankmRNA CD248801.1 - 1, 2 55 Chr. 6 - 85677082 - 85678733 [-] on Build GRCh38
GenBankmRNA CD174362.1 - 1, 2 61 Chr. 6 - 85677082 - 85678734 [-] on Build GRCh38
GenBankmRNA CD388259.1 - 1, 2 60 Chr. 6 - 85677075 - 85678732 [-] on Build GRCh38
GenBankmRNA BU536398.1 - 1, 2 40 Chr. 6 - 85677082 - 85678736 [-] on Build GRCh38
GenBankmRNA BG339050.1 - 18 Chr. - - [-] on Build
GenBankmRNA CD172704.1 - 1, 2 57 Chr. 6 - 85677082 - 85678733 [-] on Build GRCh38
GenBankmRNA BM772443.1 - 1, 2 37 Chr. 6 - 85677082 - 85678733 [-] on Build GRCh38
GenBankmRNA F25212.1 - 1, 2 20 Chr. 6 - 85677105 - 85678716 [-] on Build GRCh38
GenBankmRNA BE812115.1 - 16 Chr. 6 - 85677119 - 85678192 [-] on Build GRCh38
GenBankmRNA F24856.1 - 1, 2 16 Chr. 6 - 85677133 - 85678712 [-] on Build GRCh38
GenBankmRNA BF663376.1 - 20 Chr. - - [-] on Build
GenBankmRNA BU536630.1 - 1, 2 40 Chr. 6 - 85677079 - 85678736 [-] on Build GRCh38
GenBankmRNA CD176142.1 - 1, 2 27 Chr. 6 - 85677082 - 85678723 [-] on Build GRCh38
GenBankmRNA CD175429.1 - 1, 2 56 Chr. 6 - 85677082 - 85678731 [-] on Build GRCh38
GenBankmRNA CN368115.1 - 1, 2 34 Chr. 6 - 85677087 - 85678724 [-] on Build GRCh38
GenBankmRNA BG424974.1 - 1, 2 30 Chr. 6 - 85677084 - 85678726 [-] on Build GRCh38
GenBankmRNA BG472840.1 - 1, 2 21 Chr. 6 - 85677114 - 85678716 [-] on Build GRCh38
GenBankmRNA BX376153.2 - 1, 2 23 Chr. 6 - 85677085 - 85678719 [-] on Build GRCh38
GenBankmRNA BG760195.1 - 1, 2 20 Chr. 6 - 85677082 - 85678716 [-] on Build GRCh38
GenBankmRNA W58769.1 - 23 Chr. - - [-] on Build
GenBankmRNA BM476100.1 - 25 Chr. - - [-] on Build
GenBankmRNA CD389175.1 - 1, 2 57 Chr. 6 - 85677082 - 85678732 [-] on Build GRCh38
GenBankmRNA AJ707924.1 - 1, 2 55 Chr. 6 - 85677084 - 85678716 [-] on Build GRCh38
GenBankmRNA HY104298.1 - 1, 2 38 Chr. 6 - 85677083 - 85678732 [-] on Build GRCh38
GenBankmRNA BU600470.1 - 1, 2 43 Chr. 6 - 85677077 - 85678736 [-] on Build GRCh38
GenBankmRNA F25999.1 - 1, 2 21 Chr. 6 - 85677082 - 85678717 [-] on Build GRCh38
GenBankmRNA BM006651.1 - 1, 2 21 Chr. 6 - 85677082 - 85678716 [-] on Build GRCh38
GenBankmRNA CD638710.1 - 1, 2 40 Chr. 6 - 85677079 - 85678736 [-] on Build GRCh38
GenBankmRNA CD248379.1 - 1, 2 61 Chr. 6 - 85677082 - 85678733 [-] on Build GRCh38
GenBankmRNA CD390234.1 - 1, 2 62 Chr. 6 - 85677079 - 85678736 [-] on Build GRCh38
GenBankmRNA CD174641.1 - 1, 2 59 Chr. 6 - 85677079 - 85678733 [-] on Build GRCh38
GenBankmRNA CD246809.1 - 1, 2 53 Chr. 6 - 85677085 - 85678731 [-] on Build GRCh38
GenBankmRNA BG685417.1 - 1, 2 15 Chr. 6 - 85677082 - 85678710 [-] on Build GRCh38
GenBankmRNA CD247900.1 - 1, 2 56 Chr. 6 - 85677082 - 85678733 [-] on Build GRCh38
GenBankmRNA BG398388.1 - 15 Chr. - - [-] on Build
GenBankmRNA CD390212.1 - 1, 2 50 Chr. 6 - 85677082 - 85678733 [-] on Build GRCh38
GenBankmRNA CD000147.1 - 41 Chr. - - [-] on Build
GenBankmRNA CB110857.1 - 1, 2 37 Chr. 6 - 85677082 - 85678733 [-] on Build GRCh38
GenBankmRNA BU532819.1 - 1, 2 40 Chr. 6 - 85677082 - 85678736 [-] on Build GRCh38
GenBankmRNA BG754322.1 - 21 Chr. - - [-] on Build
GenBankmRNA BG419815.1 - 1, 2 30 Chr. 6 - 85677077 - 85678726 [-] on Build GRCh38
GenBankmRNA BG332320.1 - 1, 2 21 Chr. 6 - 85677082 - 85678716 [-] on Build GRCh38
GenBankmRNA HY103231.1 - 1, 2 35 Chr. 6 - 85677082 - 85678731 [-] on Build GRCh38
GenBankmRNA F37437.1 - 20 Chr. - - [-] on Build
GenBankmRNA HY067002.1 - 1, 2 36 Chr. 6 - 85677082 - 85678732 [-] on Build GRCh38
GenBankmRNA CD174976.1 - 1, 2 55 Chr. 6 - 85677085 - 85678733 [-] on Build GRCh38
GenBankmRNA CD387937.1 - 54 Chr. - - [-] on Build
GenBankmRNA CB117807.1 - 1, 2 36 Chr. 6 - 85677472 - 85678731 [-] on Build GRCh38
GenBankmRNA CD175900.1 - 1, 2 57 Chr. 6 - 85677079 - 85678731 [-] on Build GRCh38
GenBankmRNA BF974828.1 - 1, 2 15 Chr. 6 - 85677082 - 85678710 [-] on Build GRCh38
GenBankmRNA CN368114.1 - 1, 2 36 Chr. 6 - 85677087 - 85678720 [-] on Build GRCh38
GenBankmRNA AV712595.2 - 1, 2 37 Chr. 6 - 85677079 - 85678733 [-] on Build GRCh38
GenBankmRNA BM841447.1 - 1, 2 58 Chr. 6 - 85677134 - 85678733 [-] on Build GRCh38
GenBankmRNA CD174275.1 - 1, 2 59 Chr. 6 - 85677079 - 85678733 [-] on Build GRCh38
GenBankmRNA BG759432.1 - 1, 2 15 Chr. 6 - 85677082 - 85678710 [-] on Build GRCh38
GenBankmRNA CD173175.1 - 1, 2 53 Chr. 6 - 85677085 - 85678733 [-] on Build GRCh38
GenBankmRNA HY181073.1 - 1, 2 36 Chr. 6 - 85677083 - 85678732 [-] on Build GRCh38
GenBankmRNA AA315506.1 - 1, 2 15 Chr. 6 - 85677085 - 85678711 [-] on Build GRCh38
GenBankmRNA DW432544.1 - 1, 2 54 Chr. 6 - 85677800 - 85678736 [-] on Build GRCh38
GenBankmRNA CD174931.1 - 1, 2 54 Chr. 6 - 85677082 - 85678733 [-] on Build GRCh38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0