This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TPTEP1 SIRNA Assays ›
Gene Symbol:
TPTEP1 ERVH-1 LINC-ROR LOC101928844 TRPC7-AS2
Gene Name:
transmembrane phosphatase with tensin homology pseudogene 1
endogenous retrovirus group H member 1
long intergenic non-protein coding RNA, regulator of reprogramming
uncharacterized LOC101928844
TRPC7 antisense RNA 2
Gene Aliases:
psiTPTE22
-
ROR, lincRNA-RoR
-
CTB-28J9.2
Chromosome Location:
Chr.22: 16601911 - 16648830 on Build GRCh38
Chromosome Location:
-
Chromosome Location:
Chr.18: 57054571 - 57072119 on Build GRCh38
Chromosome Location:
Chr.19: 5847467 - 5858239 on Build GRCh38
Chromosome Location:
Chr.5: 136226655 - 136316100 on Build GRCh38
UniGene ID:
Hs.474116
Hs.383006
Hs.742426
Hs.434577
Hs.637713
Species:
Human
Species Specific ID (Flybase ID):
-
-
-
-
-
siRNA ID n269977

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TPTEP1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_001591.1 3 631
GenBank mRNA AK094889.1 - 1 1020
AY219889.1 - 3 631
EU541207.1 - 3 596
EU541208.1 - 3 596
Gene Symbol: ERVH-1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA EU669866.1 - 3 697
Gene Symbol: LINC-ROR
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_048536.1 2 398
GenBank mRNA AB844430.1 - 1 182
AB844432.1 - 1 182
AB908956.1 - 2 398
HQ315778.1 - 2 398
LC027886.1 - 2 338
LC027887.1 - 2 338
Gene Symbol: LOC101928844
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_110740.1 3 552
GenBank mRNA BC033124.1 - 3 552
Gene Symbol: TRPC7-AS2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_133682.1 2 149
GenBank mRNA BI765200.1 - 2 149
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BG333521.1 - 3 634 Chr. 22 - 16601911 - 16614237 [+] on Build GRCh38
GenBankmRNA BE299888.1 - 3 526 Chr. 19 - 5847489 - 5857902 [+] on Build GRCh38
GenBankmRNA AW572313.1 - 3 521 Chr. 22 - 16602021 - 16614178 [+] on Build GRCh38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0