This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NR2F1-AS1 SIRNA Assays ›
Gene Symbol:
NR2F1-AS1
Gene Name:
NR2F1 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.5: 93409356 - 93581320 on Build GRCh38
UniGene ID:
Hs.457407
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n269417

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: NR2F1-AS1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_021490.2 4 493
NR_021491.2 4 493
NR_109818.1 5 606
NR_109819.1 4 1168
NR_109820.1 4 493
NR_109821.1 4 493
NR_109823.1 4 490
NR_109824.1 4 493
NR_109825.1 5 1273
GenBank mRNA AK095719.1 - 5 584
AK124699.1 - 4 1218
AK311143.1 - 4 392
AK311323.1 - 4 881
BC010610.1 - 4 461
BC018089.1 - 4 470
BC042879.1 - 4 392
BC044619.1 - 5 749
BC048999.1 - 5 749
BC144330.1 - 4 1768
BC144331.1 - 4 1768
BX647510.1 - 3 261
DB257894.1 - 4 392
HY108556.1 - 4 423
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DB503335.1 - 4 389 Chr. 5 - 93544834 - 93581216 [-] on Build GRCh38
GenBankmRNA DA531679.1 - 4 544 Chr. 5 - 93543968 - 93581370 [-] on Build GRCh38
GenBankmRNA BM546601.1 - 558 Chr. - - [-] on Build
GenBankmRNA DA271332.1 - 4 407 Chr. 5 - 93438714 - 93581234 [-] on Build GRCh38
GenBankmRNA DB486010.2 - 4 389 Chr. 5 - 93411386 - 93581216 [-] on Build GRCh38
GenBankmRNA DA533837.1 - 4 466 Chr. 5 - 93544815 - 93580618 [-] on Build GRCh38
GenBankmRNA DA363045.1 - 4 537 Chr. 5 - 93553758 - 93580689 [-] on Build GRCh38
GenBankmRNA DA134670.1 - 4 392 Chr. 5 - 93411404 - 93581219 [-] on Build GRCh38
GenBankmRNA DA527197.1 - 4 476 Chr. 5 - 93411404 - 93581303 [-] on Build GRCh38
GenBankmRNA DA706940.1 - 4 475 Chr. 5 - 93544828 - 93581300 [-] on Build GRCh38
GenBankmRNA DA280798.1 - 4 374 Chr. 5 - 93411352 - 93581199 [-] on Build GRCh38
GenBankmRNA BX377086.2 - 4 396 Chr. 5 - 93543540 - 93581223 [-] on Build GRCh38
GenBankmRNA BG701915.1 - 4 636 Chr. 5 - 93501042 - 93585650 [-] on Build GRCh38
GenBankmRNA DA844094.1 - 4 472 Chr. 5 - 93544836 - 93581297 [-] on Build GRCh38
GenBankmRNA DA066475.1 - 2 157 Chr. 5 - 93411149 - 93554938 [-] on Build GRCh38
GenBankmRNA BI669922.1 - 4 397 Chr. 5 - 93411232 - 93581219 [-] on Build GRCh38
GenBankmRNA DB272535.1 - 4 494 Chr. 5 - 93544853 - 93580646 [-] on Build GRCh38
GenBankmRNA DA545967.1 - 4 392 Chr. 5 - 93544816 - 93581219 [-] on Build GRCh38
GenBankmRNA BG715542.1 - 4 477 Chr. 5 - 93411271 - 93581297 [-] on Build GRCh38
GenBankmRNA DA304607.1 - 4 379 Chr. 5 - 93411321 - 93581205 [-] on Build GRCh38
GenBankmRNA DB498974.1 - 4 389 Chr. 5 - 93544802 - 93581216 [-] on Build GRCh38
GenBankmRNA DA839566.1 - 3 283 Chr. 5 - 93432362 - 93563432 [-] on Build GRCh38
GenBankmRNA HY141502.1 - 4 373 Chr. 5 - 93544815 - 93581199 [-] on Build GRCh38
GenBankmRNA BG707159.1 - 4 397 Chr. 5 - 93411365 - 93581219 [-] on Build GRCh38
GenBankmRNA DA388648.1 - 4 543 Chr. 5 - 93544889 - 93580695 [-] on Build GRCh38
GenBankmRNA DA328801.1 - 4 394 Chr. 5 - 93543992 - 93581221 [-] on Build GRCh38
GenBankmRNA BI820847.1 - 4 443 Chr. 5 - 93342240 - 93580595 [-] on Build GRCh38
GenBankmRNA DA517345.1 - 4 469 Chr. 5 - 93411434 - 93581295 [-] on Build GRCh38
GenBankmRNA DB492381.1 - 4 389 Chr. 5 - 93544849 - 93581216 [-] on Build GRCh38
GenBankmRNA DB475119.2 - 4 467 Chr. 5 - 93411324 - 93581297 [-] on Build GRCh38
GenBankmRNA DB177914.1 - 4 473 Chr. 5 - 93438700 - 93581300 [-] on Build GRCh38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0