This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other IQSEC2 SIRNA Assays ›
Gene Symbol:
IQSEC2
Gene Name:
IQ motif and Sec7 domain 2
Gene Aliases:
BRAG1, MRX1, MRX78
Chromosome Location:
Chr.X: 53225784 - 53321350 on Build GRCh38
UniGene ID:
Hs.496138
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n269071

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: IQSEC2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243197.1 NP_001230126.1 3 600
GenBank mRNA BC038213.1 - 2 484
BC108674.1 - 3 584
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BE883792.1 - 3 376 Chr. X - 53266324 - 53281572 [-] on Build GRCh38
GenBankmRNA BQ668713.1 - 2 481 Chr. X - 53266340 - 53279659 [-] on Build GRCh38
GenBankmRNA BQ187376.1 - 1 346 Chr. X - 53266401 - 53267025 [-] on Build GRCh38
GenBankmRNA BU158163.1 - 53 Chr. - - [-] on Build
GenBankmRNA F23210.1 - 112 Chr. 2 - 110995916 - 110995931 [-] on Build GRCh38
GenBankmRNA AJ711626.1 - 1 64 Chr. X - 53266328 - 53266743 [-] on Build GRCh38

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

actin cytoskeleton organization
regulation of ARF protein signal transduction
positive regulation of GTPase activity
ARF guanyl-nucleotide exchange factor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0