This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF415 SIRNA Assays ›
Gene Symbol:
ZNF415
Gene Name:
zinc finger protein 415
Gene Aliases:
ZfLp
Chromosome Location:
Chr.19: 53107879 - 53133077 on Build GRCh38
UniGene ID:
Hs.147765
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
n268928
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF415
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NR_028343.1
5
461
XM_006723267.1
XP_006723330.1
4
393
XM_006723277.2
XP_006723340.1
4
391
XM_006723279.1
XP_006723342.1
4
550
XM_011527100.1
XP_011525402.1
4
540
XM_011527101.2
XP_011525403.1
4
515
XM_011527104.1
XP_011525406.1
4
629
XM_017026952.1
XP_016882441.1
4
343
XM_017026953.1
XP_016882442.1
4
540
XM_017026954.1
XP_016882443.1
4
417
XM_017026955.1
XP_016882444.1
4
393
XM_017026956.1
XP_016882445.1
5
543
XM_017026957.1
XP_016882446.1
4
281
XM_017026958.1
XP_016882447.1
5
597
XM_017026959.1
XP_016882448.1
4
468
XM_017026960.1
XP_016882449.1
3
183
XM_017026961.1
XP_016882450.1
4
629
XM_017026962.1
XP_016882451.1
5
572
XM_017026963.1
XP_016882452.1
4
468
XM_017026968.1
XP_016882457.1
5
656
XM_017026971.1
XP_016882460.1
4
629
XM_017026974.1
XP_016882463.1
4
507
GenBank mRNA
AK095504.1
-
5
433
BC063880.1
-
4
381
30388597
BC069668.1
-
4
380
7262632
DQ925697.1
-
4
281
DQ925698.1
-
4
481
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0