This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TTC28-AS1 SIRNA Assays ›
Gene Symbol:
TTC28-AS1
Gene Name:
TTC28 antisense RNA 1
Gene Aliases:
TTC28-AS, TTC28AS, dJ353E16.2
Chromosome Location:
Chr.22: 27919376 - 28002679 on Build GRCh38
UniGene ID:
Hs.602319
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n268352

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TTC28-AS1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026962.1 1 124
NR_026963.1 1 124
GenBank mRNA AL832597.1 - 1 79
BC016779.1 - 1 34
DA934139.1 - 1 124
DB254752.1 - 1 43
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BF243724.1 - 31 Chr. - - [-] on Build
GenBankmRNA DA172828.1 - 1 108 Chr. 22 - 27919392 - 27997631 [+] on Build GRCh38
GenBankmRNA HY096240.1 - 1 33 Chr. 22 - 27919468 - 27922404 [+] on Build GRCh38
GenBankmRNA BU858805.1 - 1 61 Chr. 22 - 27919442 - 27922154 [+] on Build GRCh38
GenBankmRNA DA121660.1 - 1 43 Chr. 22 - 27919457 - 27922461 [+] on Build GRCh38
GenBankmRNA DB444867.1 - 1 19 Chr. 22 - 27919481 - 27922380 [+] on Build GRCh38
GenBankmRNA DW434093.1 - 1 45 Chr. 22 - 27919467 - 27922178 [+] on Build GRCh38
GenBankmRNA DB187377.1 - 1 43 Chr. 22 - 27919457 - 27922484 [+] on Build GRCh38
GenBankmRNA HY066944.1 - 1 33 Chr. 22 - 27919468 - 27922395 [+] on Build GRCh38
GenBankmRNA DA870155.1 - 1 63 Chr. 22 - 27919437 - 27922446 [+] on Build GRCh38
GenBankmRNA BM921836.1 - 1 43 Chr. 22 - 27919457 - 27997616 [+] on Build GRCh38
GenBankmRNA HY077858.1 - 1 33 Chr. 22 - 27919468 - 27922411 [+] on Build GRCh38
GenBankmRNA DA126837.1 - 1 30 Chr. 22 - 27919470 - 27922519 [+] on Build GRCh38
GenBankmRNA HY084175.1 - 1 33 Chr. 22 - 27919468 - 27922414 [+] on Build GRCh38
GenBankmRNA DA779688.1 - 1 43 Chr. 22 - 27919457 - 27922518 [+] on Build GRCh38
GenBankmRNA DA779683.1 - 1 45 Chr. 22 - 27919455 - 27922516 [+] on Build GRCh38
GenBankmRNA DA503708.1 - 1 34 Chr. 22 - 27919466 - 27922485 [+] on Build GRCh38
GenBankmRNA HY172362.1 - 1 67 Chr. 22 - 27919437 - 27922366 [+] on Build GRCh38
GenBankmRNA DA124925.1 - 1 34 Chr. 22 - 27919466 - 27922496 [+] on Build GRCh38
GenBankmRNA DB074404.1 - 1 30 Chr. 22 - 27919470 - 27922485 [+] on Build GRCh38
GenBankmRNA HY029971.1 - 1 64 Chr. 22 - 27919437 - 27924525 [+] on Build GRCh38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0