siRNA Details
Gene Symbol: MIR203A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029620.1 | 1 | 38 |

More Information
Other Species Targeted


Gene Symbol: MIR203A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029620.1 | 1 | 38 |