siRNA Details
Gene Symbol: MIR31HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027054.1 | 4 | 1510 | |||
GenBank mRNA | AK124391.1 | - | 4 | 1511 | ||
BC011715.2 | - | 1 | 1025 | |||
BC021861.1 | - | 3 | 1193 |

Gene Symbol: MIR31HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027054.1 | 4 | 1510 | |||
GenBank mRNA | AK124391.1 | - | 4 | 1511 | ||
BC011715.2 | - | 1 | 1025 | |||
BC021861.1 | - | 3 | 1193 |