siRNA Details
Gene Symbol: MIR31HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027054.1 | 4 | 2027 | |||
GenBank mRNA | AK124391.1 | - | 4 | 2028 |

Gene Symbol: MIR31HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027054.1 | 4 | 2027 | |||
GenBank mRNA | AK124391.1 | - | 4 | 2028 |