This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HCG18 SIRNA Assays ›
Gene Symbol:
HCG18
Gene Name:
HLA complex group 18 (non-protein coding)
Gene Aliases:
-
Chromosome Location:
Chr.6: 30287397 - 30327156 on Build GRCh38
UniGene ID:
Hs.485041
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n265766

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 357.00
Your Price
Online offer: 296.31
296.3
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: HCG18
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024052.2 4 1258
NR_024053.2 3 1044
NR_102326.1 3 1044
GenBank mRNA AK055657.1 - 4 953
AK056160.1 - 3 262
BC035165.2 - 3 1038
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DA832767.1 - 3 349 Chr. 6 - 30294523 - 30314376 [-] on Build GRCh38
GenBankmRNA BG475820.1 - 222 Chr. - - [-] on Build
GenBankmRNA BM808048.1 - 475 Chr. - - [-] on Build
GenBankmRNA BM474246.1 - 3 438 Chr. 6 - 30294374 - 30326347 [-] on Build GRCh38
GenBankmRNA AL526171.3 - 3 464 Chr. 6 - 30292385 - 30326373 [-] on Build GRCh38
GenBankmRNA BE888347.1 - 493 Chr. - - [-] on Build
GenBankmRNA BX489584.1 - 3 273 Chr. 6 - 30294477 - 30326382 [-] on Build GRCh38
GenBankmRNA DA776066.1 - 3 551 Chr. 6 - 30294798 - 30326662 [-] on Build GRCh38
GenBankmRNA BG108155.1 - 4 501 Chr. 6 - 30294625 - 30326401 [-] on Build GRCh38
GenBankmRNA DA526907.1 - 3 477 Chr. 6 - 30294736 - 30326382 [-] on Build GRCh38
GenBankmRNA BE387468.1 - 2 262 Chr. 6 - 30292455 - 30296231 [-] on Build GRCh38
GenBankmRNA CN286179.1 - 3 439 Chr. 6 - 30294534 - 30326348 [-] on Build GRCh38
GenBankmRNA DA909557.1 - 3 365 Chr. 6 - 30294619 - 30326474 [-] on Build GRCh38
GenBankmRNA BI907017.1 - 3 476 Chr. 6 - 30294651 - 30326386 [-] on Build GRCh38
GenBankmRNA BI333149.1 - 3 556 Chr. 6 - 30294629 - 30326668 [-] on Build GRCh38
GenBankmRNA BF304181.1 - 3 269 Chr. 6 - 30292488 - 30314296 [-] on Build GRCh38
GenBankmRNA BM728475.1 - 3 451 Chr. 6 - 30294589 - 30326360 [-] on Build GRCh38
GenBankmRNA DA700189.1 - 3 262 Chr. 6 - 30294612 - 30326374 [-] on Build GRCh38
GenBankmRNA DA504712.1 - 3 474 Chr. 6 - 30294731 - 30326383 [-] on Build GRCh38
GenBankmRNA BM020076.1 - 1 23 Chr. 6 - 30292297 - 30294845 [-] on Build GRCh38
GenBankmRNA CB123757.1 - 3 324 Chr. 6 - 30294727 - 30326091 [-] on Build GRCh38
GenBankmRNA BG472090.1 - 4 671 Chr. 6 - 30294703 - 30326363 [-] on Build GRCh38
GenBankmRNA BE276734.1 - 2 259 Chr. 6 - 30292541 - 30296231 [-] on Build GRCh38
GenBankmRNA AL528846.3 - 3 455 Chr. 6 - 30292386 - 30326364 [-] on Build GRCh38
GenBankmRNA BM923783.1 - 3 468 Chr. 6 - 30292493 - 30314484 [-] on Build GRCh38
GenBankmRNA DA943926.1 - 3 530 Chr. 6 - 30294787 - 30326642 [-] on Build GRCh38
GenBankmRNA BI763370.1 - 2 235 Chr. 6 - 30292488 - 30296232 [-] on Build GRCh38
GenBankmRNA BM474248.1 - 3 436 Chr. 6 - 30294291 - 30326346 [-] on Build GRCh38
GenBankmRNA AA452964.1 - 3 287 Chr. 6 - 30294592 - 30326392 [-] on Build GRCh38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header