siRNA Details
Gene Symbol: HCG18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024052.2 | 5 | 4620 | |||
NR_024053.2 | 4 | 4406 | ||||
NR_102326.1 | 3 | 6277 |

Gene Symbol: HCG18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024052.2 | 5 | 4620 | |||
NR_024053.2 | 4 | 4406 | ||||
NR_102326.1 | 3 | 6277 |