This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SNHG17 SIRNA Assays ›
Gene Symbol:
SNHG17
Gene Name:
small nucleolar RNA host gene 17
Gene Aliases:
-
Chromosome Location:
Chr.20: 38420592 - 38435375 on Build GRCh38
UniGene ID:
Hs.400876
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n263788

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SNHG17
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015366.4 8 1032
NR_027241.2 7 1145
GenBank mRNA AK022745.1 - 4 2095
BC032119.2 - 7 1111
BC052370.1 - 7 746
BX640887.1 - 6 5140
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BM806173.1 - 3 415 Chr. 20 - 38420592 - 38426471 [-] on Build GRCh38
GenBankmRNA BM894286.1 - 1 198 Chr. 20 - 38420593 - 38420826 [-] on Build GRCh38
GenBankmRNA BM853469.1 - 4 483 Chr. 20 - 38420598 - 38431077 [-] on Build GRCh38
GenBankmRNA BF206691.1 - 6 568 Chr. 20 - 38420596 - 38434566 [-] on Build GRCh38
GenBankmRNA BG754976.1 - 6 544 Chr. 20 - 38420592 - 38434528 [-] on Build GRCh38
GenBankmRNA DB541814.1 - 3 426 Chr. 20 - 38420590 - 38426483 [-] on Build GRCh38
GenBankmRNA DW448409.1 - 115 Chr. - - [-] on Build
GenBankmRNA DW448195.1 - 115 Chr. - - [-] on Build
GenBankmRNA CV802901.1 - 3 455 Chr. 20 - 38420592 - 38426505 [-] on Build GRCh38
GenBankmRNA BG292447.1 - 6 732 Chr. 20 - 38420575 - 38435294 [-] on Build GRCh38
GenBankmRNA D20862.1 - 1 159 Chr. 20 - 38420592 - 38420792 [-] on Build GRCh38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0