This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SNHG17 SIRNA Assays ›
Gene Symbol:
SNHG17
Gene Name:
small nucleolar RNA host gene 17
Gene Aliases:
-
Chromosome Location:
Chr.20: 38420592 - 38435375 on Build GRCh38
UniGene ID:
Hs.400876
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n263787

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SNHG17
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015366.4 1 53
NR_027241.2 1 109
GenBank mRNA BC032119.2 - 1 75
BC052370.1 - 1 35
BM796603.1 - 1 62
BX640887.1 - 1 85
DA687932.1 - 1 111
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA N39481.1 - 1 46 Chr. 20 - 38426472 - 38435308 [-] on Build GRCh38
GenBankmRNA HY111703.1 - 1 63 Chr. 20 - 38421013 - 38435328 [-] on Build GRCh38
GenBankmRNA HY163163.1 - 1 63 Chr. 20 - 38434519 - 38435327 [-] on Build GRCh38
GenBankmRNA DB106805.1 - 1 90 Chr. 20 - 38422115 - 38435356 [-] on Build GRCh38
GenBankmRNA BG482037.1 - 1 66 Chr. 20 - 38420710 - 38435331 [-] on Build GRCh38
GenBankmRNA BQ880378.1 - 1 87 Chr. 20 - 38422087 - 38435353 [-] on Build GRCh38
GenBankmRNA BI117417.1 - 76 Chr. - - [-] on Build
GenBankmRNA BF311155.1 - 1 66 Chr. 20 - 38420690 - 38435331 [-] on Build GRCh38
GenBankmRNA BM720553.1 - 1 65 Chr. 20 - 38425935 - 38435331 [-] on Build GRCh38
GenBankmRNA AW328175.1 - 1 73 Chr. 20 - 38426460 - 38435338 [-] on Build GRCh38
GenBankmRNA BX508193.1 - 1 55 Chr. 20 - 38422113 - 38435320 [-] on Build GRCh38
GenBankmRNA BX508321.1 - 1 55 Chr. 20 - 38422175 - 38435320 [-] on Build GRCh38
GenBankmRNA BG481678.1 - 12 Chr. 20 - 38420597 - 38435277 [-] on Build GRCh38
GenBankmRNA BU145155.1 - 1 40 Chr. 20 - 38420641 - 38435306 [-] on Build GRCh38
GenBankmRNA BG330525.1 - 1 76 Chr. 20 - 38420726 - 38435341 [-] on Build GRCh38
GenBankmRNA BG740096.1 - 1 70 Chr. 20 - 38421034 - 38435336 [-] on Build GRCh38
GenBankmRNA DA927694.1 - 1 86 Chr. 20 - 38422243 - 38435352 [-] on Build GRCh38
GenBankmRNA HY008936.1 - 1 88 Chr. 20 - 38421065 - 38435352 [-] on Build GRCh38
GenBankmRNA BU196029.1 - 1 85 Chr. 20 - 38420644 - 38435351 [-] on Build GRCh38
GenBankmRNA BQ956982.1 - 63 Chr. - - [-] on Build
GenBankmRNA BX363714.2 - 1 37 Chr. 20 - 38420663 - 38435303 [-] on Build GRCh38
GenBankmRNA BF341505.1 - 1 75 Chr. 20 - 38422289 - 38435341 [-] on Build GRCh38
GenBankmRNA HY178299.1 - 1 112 Chr. 20 - 38426417 - 38435377 [-] on Build GRCh38
GenBankmRNA BI084345.1 - 1 52 Chr. 20 - 38420793 - 38435317 [-] on Build GRCh38
GenBankmRNA CD675826.1 - 1 44 Chr. 20 - 38420829 - 38435310 [-] on Build GRCh38
GenBankmRNA BE790426.1 - 1 15 Chr. 20 - 38420633 - 38435281 [-] on Build GRCh38
GenBankmRNA BI914409.1 - 1 92 Chr. 20 - 38422192 - 38435358 [-] on Build GRCh38
GenBankmRNA DA951419.1 - 1 88 Chr. 20 - 38426458 - 38435354 [-] on Build GRCh38
GenBankmRNA BQ673916.1 - 1 50 Chr. 20 - 38420679 - 38435316 [-] on Build GRCh38
GenBankmRNA BX332770.2 - 1 72 Chr. 20 - 38420614 - 38435338 [-] on Build GRCh38
GenBankmRNA BE394205.1 - 1 46 Chr. 20 - 38422090 - 38435312 [-] on Build GRCh38
GenBankmRNA DB233514.1 - 1 111 Chr. 20 - 38430942 - 38435377 [-] on Build GRCh38
GenBankmRNA DA959367.1 - 1 95 Chr. 20 - 38422133 - 38435360 [-] on Build GRCh38
GenBankmRNA BM456729.1 - 69 Chr. - - [-] on Build
GenBankmRNA DA948483.1 - 1 95 Chr. 20 - 38422138 - 38435360 [-] on Build GRCh38
GenBankmRNA BG106290.1 - 48 Chr. - - [-] on Build
GenBankmRNA DB286658.1 - 1 109 Chr. 20 - 38430860 - 38435375 [-] on Build GRCh38
GenBankmRNA DA087743.1 - 1 93 Chr. 20 - 38425935 - 38435359 [-] on Build GRCh38
GenBankmRNA BG481329.1 - 12 Chr. 20 - 38420625 - 38435277 [-] on Build GRCh38
GenBankmRNA BI522537.1 - 1 49 Chr. 20 - 38421006 - 38435314 [-] on Build GRCh38
GenBankmRNA BI255561.1 - 1 65 Chr. 20 - 38422152 - 38435331 [-] on Build GRCh38
GenBankmRNA BU500384.1 - 1 33 Chr. 20 - 38420792 - 38435297 [-] on Build GRCh38
GenBankmRNA CB997412.1 - 105 Chr. - - [-] on Build
GenBankmRNA DA863584.1 - 1 78 Chr. 20 - 38425825 - 38435344 [-] on Build GRCh38
GenBankmRNA DB264878.1 - 1 111 Chr. 20 - 38422110 - 38435377 [-] on Build GRCh38
GenBankmRNA DA482729.1 - 1 89 Chr. 20 - 38425927 - 38435355 [-] on Build GRCh38
GenBankmRNA DA343447.1 - 1 78 Chr. 20 - 38426451 - 38435344 [-] on Build GRCh38
GenBankmRNA DA074360.1 - 1 94 Chr. 20 - 38426461 - 38435360 [-] on Build GRCh38
GenBankmRNA BG106683.1 - 1 67 Chr. 20 - 38420653 - 38435333 [-] on Build GRCh38
GenBankmRNA DA526039.1 - 1 89 Chr. 20 - 38426464 - 38435355 [-] on Build GRCh38
GenBankmRNA BI754333.1 - 1 48 Chr. 20 - 38420592 - 38435314 [-] on Build GRCh38
GenBankmRNA BI194992.1 - 1 66 Chr. 20 - 38420746 - 38435331 [-] on Build GRCh38
GenBankmRNA BQ067601.1 - 87 Chr. - - [-] on Build
GenBankmRNA BQ669163.1 - 1 50 Chr. 20 - 38420721 - 38435316 [-] on Build GRCh38
GenBankmRNA BI195102.1 - 1 66 Chr. 20 - 38421024 - 38435331 [-] on Build GRCh38
GenBankmRNA BU153111.1 - 83 Chr. - - [-] on Build
GenBankmRNA DA582039.1 - 1 115 Chr. 20 - 38426001 - 38435381 [-] on Build GRCh38
GenBankmRNA BG292447.1 - 1 28 Chr. 20 - 38420575 - 38435294 [-] on Build GRCh38
GenBankmRNA DA959431.1 - 1 78 Chr. 20 - 38422120 - 38435344 [-] on Build GRCh38
GenBankmRNA BG336608.1 - 83 Chr. - - [-] on Build

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0