This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SNHG17 SIRNA Assays ›
Gene Symbol:
SNHG17
Gene Name:
small nucleolar RNA host gene 17
Gene Aliases:
-
Chromosome Location:
Chr.20: 38420592 - 38435375 on Build GRCh38
UniGene ID:
Hs.400876
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n263784

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SNHG17
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015366.4 8 879
NR_027241.2 7 992
GenBank mRNA AK022745.1 - 4 1942
BC032119.2 - 7 958
BC052370.1 - 7 593
BX640887.1 - 6 4987
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BM806173.1 - 3 262 Chr. 20 - 38420592 - 38426471 [-] on Build GRCh38
GenBankmRNA BM853469.1 - 4 330 Chr. 20 - 38420598 - 38431077 [-] on Build GRCh38
GenBankmRNA BF206691.1 - 6 415 Chr. 20 - 38420596 - 38434566 [-] on Build GRCh38
GenBankmRNA BF340988.1 - 7 591 Chr. 20 - 38420709 - 38435274 [-] on Build GRCh38
GenBankmRNA BX332770.2 - 8 776 Chr. 20 - 38420614 - 38435338 [-] on Build GRCh38
GenBankmRNA CV802901.1 - 3 302 Chr. 20 - 38420592 - 38426505 [-] on Build GRCh38
GenBankmRNA BF311155.1 - 7 609 Chr. 20 - 38420690 - 38435331 [-] on Build GRCh38
GenBankmRNA BG481329.1 - 7 689 Chr. 20 - 38420625 - 38435277 [-] on Build GRCh38
GenBankmRNA BU145155.1 - 6 598 Chr. 20 - 38420641 - 38435306 [-] on Build GRCh38
GenBankmRNA BM894286.1 - 1 45 Chr. 20 - 38420593 - 38420826 [-] on Build GRCh38
GenBankmRNA BG754976.1 - 6 391 Chr. 20 - 38420592 - 38434528 [-] on Build GRCh38
GenBankmRNA BG106683.1 - 6 623 Chr. 20 - 38420653 - 38435333 [-] on Build GRCh38
GenBankmRNA DB541814.1 - 3 273 Chr. 20 - 38420590 - 38426483 [-] on Build GRCh38
GenBankmRNA BU196029.1 - 6 643 Chr. 20 - 38420644 - 38435351 [-] on Build GRCh38
GenBankmRNA BI754333.1 - 6 604 Chr. 20 - 38420592 - 38435314 [-] on Build GRCh38
GenBankmRNA BX363714.2 - 8 863 Chr. 20 - 38420663 - 38435303 [-] on Build GRCh38
GenBankmRNA BI194992.1 - 7 611 Chr. 20 - 38420746 - 38435331 [-] on Build GRCh38
GenBankmRNA EL953190.1 - 6 587 Chr. 20 - 38420773 - 38434566 [-] on Build GRCh38
GenBankmRNA BG292447.1 - 6 584 Chr. 20 - 38420575 - 38435294 [-] on Build GRCh38
GenBankmRNA BE790426.1 - 7 572 Chr. 20 - 38420633 - 38435281 [-] on Build GRCh38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0