This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LINC01139 SIRNA Assays ›
Gene Symbol:
LINC01139
Gene Name:
long intergenic non-protein coding RNA 1139
Gene Aliases:
-
Chromosome Location:
Chr.1: 238480384 - 238486017 on Build GRCh38
UniGene ID:
Hs.532047
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n263680

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LINC01139
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015407.1 2 687
GenBank mRNA BC044843.1 - 2 687
BC064529.1 - 2 693
BC073894.1 - 2 664
BC113688.1 - 1 403
BC113690.1 - 1 403
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BI761548.1 - 2 686 Chr. 1 - 238481108 - 238486017 [-] on Build GRCh38
GenBankmRNA CD299975.1 - 2 666 Chr. 1 - 238481187 - 238485997 [-] on Build GRCh38
GenBankmRNA BC016627.1 - 1 1694 Chr. 9 - 69084981 - 69087181 [+] on Build GRCh38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0