This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LINC01139 SIRNA Assays ›
Gene Symbol:
LINC01139
Gene Name:
long intergenic non-protein coding RNA 1139
Gene Aliases:
-
Chromosome Location:
Chr.1: 238480384 - 238486017 on Build GRCh38
UniGene ID:
Hs.532047
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n263679

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LINC01139
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015407.1 2 374
GenBank mRNA BC044843.1 - 2 374
BC064529.1 - 2 380
BC073894.1 - 2 351
BC113688.1 - 1 90
BC113690.1 - 1 90
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BI761548.1 - 2 373 Chr. 1 - 238481108 - 238486017 [-] on Build GRCh38
GenBankmRNA BF695650.1 - 2 363 Chr. 1 - 238481218 - 238486004 [-] on Build GRCh38
GenBankmRNA BM468986.1 - 2 391 Chr. 1 - 238481089 - 238486023 [-] on Build GRCh38
GenBankmRNA BG532901.1 - 2 393 Chr. 1 - 238481174 - 238486035 [-] on Build GRCh38
GenBankmRNA BF696789.1 - 355 Chr. - - [-] on Build
GenBankmRNA CD299975.1 - 2 354 Chr. 1 - 238481187 - 238485997 [-] on Build GRCh38
GenBankmRNA BX341941.2 - 2 381 Chr. 1 - 238481162 - 238486024 [-] on Build GRCh38
GenBankmRNA BG612440.1 - 2 391 Chr. 1 - 238481204 - 238486032 [-] on Build GRCh38
GenBankmRNA BG429941.1 - 2 357 Chr. 1 - 238481191 - 238486001 [-] on Build GRCh38
GenBankmRNA DN992082.1 - 2 389 Chr. 1 - 238481244 - 238486027 [-] on Build GRCh38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0