This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LINC01106 SIRNA Assays ›
Gene Symbol:
LINC01106 LINC01123
Gene Name:
long intergenic non-protein coding RNA 1106
long intergenic non-protein coding RNA 1123
Gene Aliases:
-
-
Chromosome Location:
Chr.2: 110375109 - 110384536 on Build GRCh38
Chromosome Location:
Chr.2: 109986980 - 109995281 on Build GRCh38
UniGene ID:
Hs.643369
Hs.516403
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID n263537

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LINC01106
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027244.1 2 260
GenBank mRNA AK095678.1 - 2 569
BC101449.1 - 2 568
BC101450.1 - 2 568
BC101451.1 - 2 568
BC113052.1 - 2 568
BP341570.1 - 2 246
CR749351.1 - 3 351
Gene Symbol: LINC01123
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_046110.1 4 1676
NR_046111.1 3 1491
NR_046112.1 2 249
GenBank mRNA AK056084.1 - 4 1593
BC101448.1 - 2 569
DA999847.1 - 2 180
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AW675296.1 - 2 243 Chr. 2 - 109986987 - 109994593 [+] on Build GRCh38
GenBankmRNA BI821206.1 - 2 136 Chr. 2 - 109987093 - 109994923 [+] on Build GRCh38
GenBankmRNA BP199484.1 - 2 166 Chr. 2 - 109987063 - 109994857 [+] on Build GRCh38
GenBankmRNA BI914615.1 - 2 153 Chr. 2 - 109987078 - 109995113 [+] on Build GRCh38
GenBankmRNA HY149276.1 - 2 167 Chr. 2 - 109987063 - 109994745 [+] on Build GRCh38
GenBankmRNA BE167578.1 - 2 235 Chr. 2 - 109986996 - 109994775 [+] on Build GRCh38

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0